missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANGPTL5, Mouse anti-Human, Clone: 1B2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00253935-M01/DEL
This item is not returnable.
View return policy
Description
Sequence: MMSPSQASLLFLNVCIFICGEAVQGNCVHHSTDSSVVNIVEDGSNAKDESKSNDTVCKEDCEESCDVKTKITREEKHFMCRNLQNSIVSYTRSTKKLLRNMMDEQQASLDYLSNQVNELMNRVLLLTTEVFRKQLDPFPHRPVQSHGLDCTDIKDTIGSVTKTPSGLYIIHPEGSSYPFEVMCDMDYRGGGWTVIQKRIDGIIDFQRLWCDYLDGFGDLLGEFWLGLKKIFYIVNQKNTSFMLYVALESEDDTLAYASYDNFWLEDETRFFKMHLGRYSGNAGDAFRGLKREDNQNAKPFSTSDVDNDGCRPACLVNGQSVKSCSHLHNKTGWWFNECGLANLNGIHHFSGKLLATGIQWGTWTKNNSPVKIKSVSMKIRRMYNPYFKSpecifications
| ANGPTL5 | |
| Monoclonal | |
| Unconjugated | |
| angiopoietin-like 5 | |
| - | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunoprecipitation, Western Blot | |
| 1B2 | |
| In 1x PBS, pH 7.4 | |
| BC049170 | |
| ANGPTL5 | |
| ANGPTL5 (AAH49170.1, 1 a.a. ∼ 388 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 253935 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction