missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGPTL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103605
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ANGPTL5.
Predicted to be active in collagen-containing extracellular matrix and extracellular space.
Specifications
| ANGPTL5 | |
| Polyclonal | |
| Unconjugated | |
| ANGPTL5 | |
| angiopoietin like 5; angiopoietin-like 5; angiopoietin-like protein 5; Angiopoietin-related protein 5; ANGPTL5; UNQ5795/PRO19600 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 253935 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86XS5 | |
| ANGPTL5 | |
| A synthesized peptide derived from human ANGPTL5(Accession Q86XS5), corresponding to amino acid residues R277-L327. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction