missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANGPTL6 Monoclonal Antibody (766544)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA524370
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
May play a role in the wound healing process. May promote epidermal proliferation, remodeling and regeneration. May promote the chemotactic activity of endothelial cells and induce neovascularization. May counteract high-fat diet-induced obesity and related insulin resistance through increased energy expenditure.
Specifications
| ANGPTL6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8R0Z6 | |
| ANGPTL6 | |
| E. coli-derived recombinant mouse Angiopoietin-like Protein 6/ANGPTL6 Ala25-Leu457. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 766544 | |
| Unconjugated | |
| ANGPTL6 | |
| 6330404E11Rik; AGF; angiopoietin like 6; angiopoietin-like 6; angiopoietin-like protein 6; Angiopoietin-related growth factor; angiopoietin-related protein 5; Angiopoietin-related protein 6; Angptl6; ARP3; Arp5; UNQ152/PRO178 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 70726 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction