missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANGPTL7, Mouse anti-Human, Clone: 1A2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00010218-M06.100ug
This item is not returnable.
View return policy
Description
Sequence: QKLSKHKTPAQPQLKAANCCEEVKELKAQVANLSSLLSELNKKQERDWVSVVMQVMELESNSKRMESRLTDAESKYSEMNNQIDIMQLQAAQTVTQTSADSpecifications
| ANGPTL7 | |
| Monoclonal | |
| Unconjugated | |
| angiopoietin-like 7 | |
| AngX/CDT6/RP4-647M16.2/dJ647M16.1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1A2 | |
| In 1x PBS, pH 7.4 | |
| NM_021146 | |
| ANGPTL7 | |
| ANGPTL7 (NP_066969, 27 a.a. ∼ 126 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10218 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction