Learn More
Invitrogen™ ANGPTL7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113900
Description
Immunogen sequence: NQIDIMQLQA AQTVTQTSAD AIYDCSSLYQ KNYRISGVYK LPPDDFLGSP For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.
Angptl7 (angiopoietin-like 7), alternately known as cornea-derived transcript 6 protein (CDT6) or AngX, is a 346 amino acid secreted protein that is expressed specifically in the stromal layer of cornea. Angptl7 forms disulfide-linked homotetramers via coiled-coil interactions and contains one fibrinogen C-terminal domain. Overexpressed in patients with glaucoma, Angptl7 serves as a potential therapeutic target for glaucoma and has been found to increase collagen expression. The gene encoding Angptl7 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome.
Specifications
| ANGPTL7 | |
| Polyclonal | |
| Unconjugated | |
| ANGPTL7 | |
| angiopoietin like 7; angiopoietin-like 7; Angiopoietin-like factor; angiopoietin-like factor (CDT6); angiopoietin-like protein 7; Angiopoietin-related protein 7; ANGPTL7; AngX; CDT6; Cornea-derived transcript 6 protein; dJ647M16.1; UNQ313/PRO356 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10218 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O43827 | |
| ANGPTL7 | |
| Synthetic peptide directed towards the middle region of human ANGPTL7. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.