Learn More
Aniline sulfate, 97%
Product Code. p-7022676
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
Product Code. 11453017
Brand: Thermo Scientific Alfa Aesar A15563.22
Request Bulk or Custom Format
CAS: 542-16-5 | (C6H5NH2)2·H2SO4
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 542-16-5
Molecular Formula (C6H5NH2)2·H2SO4
MDL Number MFCD00013110
Synonym Benzenamine sulfate 2:1
Specifications
Density 1.377
Odor Odorless
Quantity 100 g
UN Number UN2811
Beilstein 3729533
Solubility Information Soluble in water
Formula Weight 284.34
Percent Purity 98%
Chemical Name or Material Aniline sulfate