missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anillin Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-41099
This item is not returnable.
View return policy
Description
Anillin Polyclonal specifically detects Anillin in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Anillin | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| actin-binding protein anillin, ANILLIN, anillin (Drosophila Scraps homolog), actin binding protein, anillin, actin binding protein, anillin, actin binding protein (scraps homolog, Drosophila), DKFZp779A055, Drosophila Scraps homolog, scra, Scraps | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 54443 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_061155.2 | |
| ANLN | |
| Peptide with sequence C-TYPDDEKRKNP corresponding to internal region according to NP_061155.2. | |
| 0.1 mg | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction