missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios™ Aniosept 41 Premium

Product Code. 15378944
Change view
Click to view available options
Volume (Metric):
400 mL
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15378944 400 mL Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15378944 Supplier Laboratoires Anios™ Supplier No. 2466330

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Disinfection of surfaces and equipments for medical, paramedical department, etc. Disinfection of furnitures, bedding, chair, etc. Atmosphere deodorizing. Apropriate for waiting room.

DÉSODORISANT D’ATMOSPHÈRE / DÉSINFECTANT
Large spectre d’activité :

  • Bactéricide, fongicide, actif sur virus.
  • Séchage rapide.
  • Parfum agréable.
  • Pulvérisation rapide, uniforme et fine.

Specifications

Product Type Aniosept 41 Premium
Quantity 12/Pk.
Volume (Metric) 400 mL
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable and non-flammable aerosols category 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H222-Extremely flammable aerosol.
  • H229-Pressurised container: May burst if heated.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P211-Do not spray on an open flame or other ignition source.
  • P251-Do not pierce or burn, even after use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ethanol, N-(3--aminopropane-1,3-D amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.