missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANK2 Mouse anti-Human, Biotin, Clone:S105-13, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human ANK2.
Brand: Abnova MAB16997.100ug
This item is not returnable.
View return policy
Specifications
| ANK2 | |
| Monoclonal | |
| Biotin | |
| Immunocytochemistry (1:200) Immunofluorescence (1:200) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| ANK2 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S105-13 | |
| ankyrin 2, neuronal | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| DKFZp686M09125, DKFZp686P0948, FLJ38277, LQT4 | |
| A synthetic peptide corresponding to amino acids 203-496 of human ANK2. | |
| 287 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction