Learn More
ANKLE2 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41145
 View more versions of this product
Product Code. 18160199
  • € 507.00 / 0.10mg
Description
ANKLE2 Polyclonal specifically detects ANKLE2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ANKLE2
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
ANKLE2
Antibody was raised against a 19 amino acid synthetic peptide near the center of human ANKLE2. The immunogen is located within amino acids 280 - 330 of ANKLE2. Amino Acid Squence: CSESETVNKERANSYKNPRT
Affinity Purified
RUO
23141
Human, Mouse, Rat
IgY
Western Blot, ELISA
1 mg/mL
Western Blot, ELISA
ankyrin repeat and LEM domain containing 2, ankyrin repeat and LEM domain-containing protein 2, FLJ22280, FLJ36132, KIAA0692, LEM domain containing 7, LEMD7
Chicken
121 kDa
0.1 mg
Primary
Three isoforms of ANKLE2 are known to exist; this antibody will detect all three. ANKLE2 antibody is predicted to not cross-react with ANKLE1.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.