missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKLE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109389
This item is not returnable.
View return policy
Description
Immunogen sequence: CRYNVMHVAA KENQASICQL TLDVLENPDF MRLMYPDDDE AMLQKRIRYV VDLYLNTPDK MGYDTPLHFA CKFGNADVVN VLSSHHLIVK NSRNKYDKTP E.
This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined.
Specifications
| ANKLE2 | |
| Polyclonal | |
| Unconjugated | |
| ANKLE2 | |
| 1110001J12Rik; Ab2-034; AI661024; Ankle2; ankyrin repeat and LEM domain containing 2; ankyrin repeat and LEM domain-containing protein 2; D5Ertd585e; Kiaa0692; LEM domain containing 7; LEM domain-containing protein 4; LEM4; LEMD7; liver regeneration-related protein LRRG057; MCPH16; RGD1310191 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23141, 360829, 71782 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.05 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6P1H6, Q7TP65, Q86XL3 | |
| ANKLE2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-450 of human ANKLE2 (NP_055929.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction