missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRA2 Antibody (OTI8A9), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72105AF532
This item is not returnable.
View return policy
Description
ANKRA2 Monoclonal specifically detects ANKRA2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ANKRA2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ANKRA2 | |
| Full length recombinant protein of human ANKRA2 (NP_075526) produced in E.coli. | |
| 0.1 mL | |
| 57763.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI8A9 | |
| Western Blot | |
| ANKRARFXANK-like protein 2, ankyrin repeat family A protein 2, ankyrin repeat, family A (RFXANK-like), 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction