missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRA2 Antibody (OTI8A9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72105F
This item is not returnable.
View return policy
Description
ANKRA2 Monoclonal antibody specifically detects ANKRA2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ANKRA2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 57763 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI8A9 | |
| Western Blot | |
| ANKRARFXANK-like protein 2, ankyrin repeat family A protein 2, ankyrin repeat, family A (RFXANK-like), 2 | |
| Full length human recombinant protein of human ANKRA2 (NP_075526) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction