missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRA2 Monoclonal Antibody (OTI8A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808378S
This item is not returnable.
View return policy
Description
ANKRA2 Monoclonal antibody specifically detects ANKRA2 in Human samples. It is validated for Western BlotSpecifications
| ANKRA2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86YS3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84440 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8A9 | |
| Unconjugated | |
| RAB11FIP4 | |
| 1110004M18Rik; ANKRA; ANKRA2; ankyrin repeat family A member 2; ankyrin repeat family A protein 2; ankyrin repeat family A protein 2-like protein; ankyrin repeat, family A (RFXANK-like), 2; ankyrin-repeat family A protein; RFXANK-like protein 2; similar to SLO-interacting ankyrin-containing protein; SLO-interacting ankyrin-containing protein; wu:fd08c02; zgc:64138 | |
| Human recombinant protein fragment corresponding to amino acids 91-177 of human RAB11FIP4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction