missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANKRA2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15818123
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15818123 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15818123 Supplier Invitrogen™ Supplier No. PA569710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 75%; Zebrafish: 100%.

This protein may facilitate endocytosis by linking megalin to components of the cytoskeleton or endocytic machinery. ANKRA2 is a megalin- and BKCa potassium channel-interacting factor. It is a protein known to interact with HDAC4 and HDAC5.ANKRA2 is important for transcriptional repression by AhRR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRA2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANKRA2
Gene Accession No. Q99PE2
Gene Alias 1110004M18Rik; ANKRA; ANKRA2; ankyrin repeat family A member 2; ankyrin repeat family A protein 2; ankyrin repeat family A protein 2-like protein; ankyrin repeat, family A (RFXANK-like), 2; ankyrin-repeat family A protein; RFXANK-like protein 2; similar to SLO-interacting ankyrin-containing protein; SLO-interacting ankyrin-containing protein; wu:fd08c02; zgc:64138
Gene Symbols ANKRA2
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of mouse ANKRA2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68558
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.