missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD13A Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18496CL1
This item is not returnable.
View return policy
Description
ANKRD13A Polyclonal antibody specifically detects ANKRD13A in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ANKRD13A | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 88455 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| ANKRD13, ankyrin repeat domain 13, ankyrin repeat domain 13A, ankyrin repeat domain-containing protein 13A, NY-REN-25, NY-REN-25 antigen, Protein KE03 | |
| Recognizes region between residues Between between 500 and 550.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction