missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD13A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23998-1-AP-20UL
This item is not returnable.
View return policy
Description
ANKRD13A, also named as ANKRD13, a 590 amino acid protein, which contains two ANK repeats and four UIM (ubiquitin-interacting motif) repeats. ANKRD13A localizes in the cell membrane. ANKRD13A as ubiquitin-binding protein that specifically recognizes and binds ′Lys-63′-linked ubiquitin, not bind ′Lys-48′-linked ubiquitin. ANKRD13A positively regulates the internalization of ligand-activated EGFR by binding to the Ub moiety of ubiquitinated EGFR at the cell membrane.Specifications
| ANKRD13A | |
| Polyclonal | |
| Unconjugated | |
| ANKRD13A | |
| ANKRD13, ANKRD13A, ankyrin repeat domain 13A, NY REN 25, Protein KE03 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 88455 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IZ07 | |
| ANKRD13A | |
| ANKRD13A Fusion Protein Ag21200 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction