missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD13B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25737-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody specifically recognizes the 70 kDa protein.
ANKRD13B (ankyrin repeat domain 13B) is an ubiquitin-binding protein belongs to the Ankrd 13 family. It specifically recognizes and binds ′Lys-63′-linked ubiquitin, and positively regulates the internalization of ligand-activated EGFR by binding to the Ub moiety of ubiquitinated EGFR at the cell membrane.Specifications
| ANKRD13B | |
| Polyclonal | |
| Unconjugated | |
| ANKRD13B | |
| ANKRD13B, ankyrin repeat domain 13B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 124930, 268445 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5F259, Q86YJ7 | |
| ANKRD13B | |
| ANKRD13B Fusion Protein Ag22732 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction