missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD15 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81953AF700
This item is not returnable.
View return policy
Description
ANKRD15 Polyclonal antibody specifically detects ANKRD15 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ANKRD15 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| ANKRD15ankyrin repeat domain 15, Ankyrin repeat domain-containing protein 15, KANKDKFZp451G231, KIAA0172MGC43128, Kidney ankyrin repeat-containing protein, KN motif and ankyrin repeat domain-containing protein 1, KN motif and ankyrin repeat domains 1 | |
| ANKRD15 antibody was raised against a 19 amino acid peptide near the amino terminus of human ANKRD15 . The immunogen is located within the first 50 amino acids of ANKRD15. Amino Acid Squence: DILSGDQDKEQKDPYFVET | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23189 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction