missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD15 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03345UV
This item is not returnable.
View return policy
Description
ANKRD15 Polyclonal antibody specifically detects ANKRD15 in Human samples. It is validated for Western Blot
Specifications
| ANKRD15 | |
| Polyclonal | |
| Western Blot | |
| ANKRD15ankyrin repeat domain 15, Ankyrin repeat domain-containing protein 15, KANKDKFZp451G231, KIAA0172MGC43128, Kidney ankyrin repeat-containing protein, KN motif and ankyrin repeat domain-containing protein 1, KN motif and ankyrin repeat domains 1 | |
| The immunogen recognized by this antibody maps to a region between residue 1302 and 1352 of human KN motif and ankyrin repeat domains 1 using the numbering given in entry NP_055973.2 (GeneID 23189). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23189 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction