missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD16 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24334-1-AP-150UL
This item is not returnable.
View return policy
Description
Required to prevent the mis activation of serine (Ser) with tRNA(Ala) by promoting the hydrolysis of Ser-mischarged tRNA(Ala), thereby playing a role in translational fidelity. Binds directly to the catalytic domain of AARS/AlaRS and captures Ser that is mis activated by AARS/AlaRS, preventing the charging of Ser adenylates to tRNA(Ala) and precluding Ser misincorporation in nascent peptides.Specifications
| ANKRD16 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD16 | |
| ANKRD16, ankyrin repeat domain 16 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 320816, 54522 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A2AS55, Q6P6B7 | |
| Ankrd16 | |
| ANKRD16 Fusion Protein Ag19369 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction