missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANKRD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17157323
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17157323 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17157323 Supplier Invitrogen™ Supplier No. PA5109391

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GTMEDSEAVQ RATALIEQRL AQEEENEKLR GDARQKLPMD LLVLEDEKHH GAQSAALQKV KGQERVRKTS LDLRREIIDV GGIQNLIELR KKRKQKKRDA LAASHEPPPE P.

ANKRD2 may play an important role in skeletal muscle hypertrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ankrd2
Gene Accession No. Q9WV06
Gene Alias Ankrd2; ankyrin repeat domain 2; ankyrin repeat domain 2 (stretch responsive muscle); ankyrin repeat domain-containing protein 2; ankyrin repeat domain-containing protein 2; LOW QUALITY PROTEIN: ankyrin repeat domain-containing protein 2; ARPP; hArpp; LOW QUALITY PROTEIN: ankyrin repeat domain-containing protein 2; mArpp; Skeletal muscle ankyrin repeat protein
Gene Symbols Ankrd2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-140 of human ANKRD2 (NP_001123453.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309374, 56642
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.