missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANKRD22 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15960515
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15960515 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15960515 Supplier Invitrogen™ Supplier No. PA571839

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANKRD22 Polyclonal Antibody for Western Blot, IHC (P)

ANKRD22 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD22
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ANKRD22
Gene Accession No. Q5VYY1, Q9D3J5
Gene Alias 5430429D21Rik; Ankrd22; ankyrin repeat domain 22; ankyrin repeat domain-containing protein 22; D19Ertd675e
Gene Symbols ANKRD22
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 140-169 amino acids from the C-terminal region of human ANKRD22.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118932, 52024
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.