missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559240
This item is not returnable.
View return policy
Description
Immunogen sequence: DNAKLKVTVK KQMDKIEELQ KNLLNANLSE DEKEQLKKLM ELKQSLECNL DQEMKKNVEL EREITGFKNL Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 56%.
ANKRD26 encodes a protein containing N-terminal ankyrin repeats which function in protein-protein interactions. Mutations in this gene are associated with autosomal dominant thrombocytopenia-2. Pseudogenes of this gene are found on chromosome 7, 10, 13 and 16. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| ANKRD26 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD26 | |
| 5730521P14Rik; ANKRD26; ankyrin repeat domain 26; ankyrin repeat domain-containing protein 26; bA145E8.1; BC008111; KIAA1074; mKIAA1074; RGD1310736; THC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22852 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UPS8 | |
| ANKRD26 | |
| Recombinant protein corresponding to Human ANKRD26. Recombinant protein control fragment (Product #RP-97038). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction