missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD27 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50029AF405
This item is not returnable.
View return policy
Description
ANKRD27 Polyclonal antibody specifically detects ANKRD27 in Human samples. It is validated for Western Blot
Specifications
| ANKRD27 | |
| Polyclonal | |
| Western Blot | |
| ankyrin repeat domain 27 (VPS9 domain), ankyrin repeat domain-containing protein 27, DKFZp434L0718, FLJ00040, VARP, Vps9 domain and ankyrin-repeat-containing protein, VPS9 domain-containing protein, VPS9-ankyrin-repeat protein | |
| The immunogen for this product maps to a region between residue 225 and 275 of human Vps9 Domain and Ankyrin-Repeat-Containing Protein using the numbering given in entry NP_115515.2 (GeneID 84079). | |
| 0.1 mL | |
| Signal Transduction | |
| 84079 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction