missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD30BL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145130
This item is not returnable.
View return policy
Description
Immunogen sequence: MERLSAAPVK GQTGPERPSP FSQLVYTNND SYVIHHGDLR KIHKA Antigen sequence identity: Mouse 40%; Rat 40%.
ANKRD30BL is a protein that belongs to the ankyrin repeat domain-containing family. It is expressed in various tissues and has been suggested to play a role in cell proliferation and differentiation. However, its exact function is still not fully understood.
Specifications
| ANKRD30BL | |
| Polyclonal | |
| Unconjugated | |
| ANKRD30BL | |
| ANKRD30BL | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 1-45. Recombinant protein control fragment (Product #RP-109803). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A7E2S9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 554226 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction