missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566099
This item is not returnable.
View return policy
Description
Immunogen sequence: NGMQDSRKKLD QMRSQFQEIQ DQLTATIRCT KEMEGD Highest antigen sequence identity to the following orthologs - mouse 38%, rat 35%.
ANKRD36 (Ankyrin Repeat Domain 36) is a Protein Coding gene. An important paralog of this gene isANKRD36C.
Specifications
| ANKRD36 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD36 | |
| ANKRD36; ankyrin repeat domain 36; ankyrin repeat domain-containing protein 36; ankyrin repeat domain-containing protein 36A; ankyrin-related; UNQ2430 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 375248 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6QL64 | |
| ANKRD36 | |
| Recombinant Human ANKRD36. Recombinant protein control fragment (Product #RP-105631). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction