missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566135
This item is not returnable.
View return policy
Description
Immunogen sequence: ASVNAPPDPCK QSPVHLAAGS GLACFLLWQL QTGADLNQQD VLGEAPLHKA AKVGSLECLS LLV Highest antigen sequence identity to the following orthologs - mouse 91%, rat 94%.
ANKRD37 (Ankyrin Repeat Domain 37) is a Protein Coding gene. An important paralog of this gene isANKRD42.
Specifications
| ANKRD37 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD37 | |
| Ankrd37; ankyrin repeat domain 37; ankyrin repeat domain-containing protein 37; hLrp2bp; low density lipoprotein receptor-related protein binding protein; low-density lipoprotein receptor-related protein 2-binding protein; LPR2BP; Lrp2bp; RGD1565715 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 353322 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z713 | |
| ANKRD37 | |
| Recombinant Human ANKRD37. Recombinant protein control fragment (Product #RP-106820). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction