missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD50 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50031AF350
This item is not returnable.
View return policy
Description
ANKRD50 Polyclonal antibody specifically detects ANKRD50 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ANKRD50 | |
| Polyclonal | |
| Western Blot | |
| ankyrin repeat domain 50 | |
| The immunogen for this product maps to a region between residue 1200 and 1250 of human Ankyrin Repeat Domain 50 using the numbering given in entry NP_065070.1 (GeneID 57182). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 57182 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction