missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD53 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15400
This item is not returnable.
View return policy
Description
ANKRD53 Polyclonal specifically detects ANKRD53 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ANKRD53 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
| ANKRD53 | |
| Recombinant protein encompassing a sequence within the center region of human ANKRD53. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 79998 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| ankyrin repeat domain 53, ankyrin repeat domain-containing protein 53, FLJ12056, FLJ36160 | |
| Rabbit | |
| 38 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction