missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD53 Antibody (OTI1E1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72217AF488
This item is not returnable.
View return policy
Description
ANKRD53 Monoclonal specifically detects ANKRD53 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ANKRD53 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ANKRD53 | |
| Recombinant protein fragment corresponding to amino acids 1-300 of human ANKRD53 (NP_079209) produced in E.coli. | |
| 0.1 mL | |
| 79998.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1E1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ankyrin repeat domain 53, ankyrin repeat domain-containing protein 53, FLJ12056, FLJ36160 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction