missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD53 Monoclonal Antibody (OTI1E1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525470
This item is not returnable.
View return policy
Description
ANKRD53 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Required for normal progression through mitosis. Involved in chromosome alignment and cytokinesis via regulation of microtubules polymerization.
Specifications
| ANKRD53 | |
| Monoclonal | |
| 0.58 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N9V6 | |
| ANKRD53 | |
| Human recombinant protein fragment corresponding to amino acids 1-300 of ANKRD53 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1E1 | |
| Unconjugated | |
| ANKRD53 | |
| 4930564N15Rik; ANKRD53; ankyrin repeat domain 53; ankyrin repeat domain-containing protein 53 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79998 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction