missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD54 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | ANKRD54 |
|---|---|
| Clone | 1E9A9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234093
|
Proteintech
68388-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236418
|
Proteintech
68388-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ANKRD54 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ANKRD54. This antibody recognizes human, mouse, rat antigen. The ANKRD54 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| ANKRD54 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 129138 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
| 1E9A9 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title