missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD65 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564240
This item is not returnable.
View return policy
Description
Immunogen sequence: EEEQELRWME LDSEEALGTR TEGPSVVQGW GHLLQAVWRG PAGLVTQLLR QGH Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 51%.
The function remain unknown.
Specifications
| ANKRD65 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD65 | |
| ANKRD65; ankyrin repeat domain 65; ankyrin repeat domain-containing protein 65; hCG_20426; LOC441869 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 441869 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| E5RJM6 | |
| ANKRD65 | |
| Recombinant protein corresponding to Human ANKRD65. Recombinant protein control fragment (Product #RP-103609). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction