missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANKRD9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15838293
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15838293 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15838293 Supplier Invitrogen™ Supplier No. PA569883

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.

ANKRD9 gene ontology annotations related to this gene include post-translational protein modification; protein localization to plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ANKRD9
Gene Accession No. Q8BH83
Gene Alias 2500003O20Rik; Ankrd9; ankyrin repeat domain 9; ankyrin repeat domain-containing protein 9
Gene Symbols ANKRD9
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ANKRD9 (aa 270-319).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74251
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.