missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKS1A Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71806DL594
This item is not returnable.
View return policy
Description
ANKS1A Polyclonal antibody specifically detects ANKS1A in Human samples. It is validated for Western Blot
Specifications
| ANKS1A | |
| Polyclonal | |
| Western Blot | |
| ANKS1ankyrin repeat and SAM domain-containing protein 1A, ankyrin repeat and SAM domain containing 1, ankyrin repeat and sterile alpha motif domain containing 1, ankyrin repeat and sterile alpha motif domain containing 1A, KIAA0229MGC42354, ODIN | |
| The immunogen for this product maps to a region between residue 875 and 925 of human Ankyrin Repeat and Sterile Apha Motif Domain Containing 1A using the numbering given in entry NP_056060.2 (GeneID 23294). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23294 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction