missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKS1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545780
This item is not returnable.
View return policy
Description
Peptide sequence: ACAKMRANCQ KSTEQMKKVP TIILSVSYKG VKFIDATNKN IIAEHEIRNI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes a multi-domain protein that is predominantly expressed in brain and testis. This protein interacts with amyloid beta protein precursor (AbetaPP) and may have a role in normal brain development, and in the pathogenesis of Alzheimer's disease. Expression of this gene has been shown to be elevated in patients with pre-B cell acute lymphocytic leukemia associated with t(1;19) translocation. Alternatively spliced transcript variants encoding different isoforms (some with different subcellular localization) have been described for this gene.
Specifications
| ANKS1B | |
| Polyclonal | |
| Unconjugated | |
| ANKS1B | |
| AIDA; AIDA-1; AIDA-1b; amyloid-beta precursor protein intracellular domain associated protein 1; amyloid-beta protein intracellular domain-associated protein 1; ANKS1B; ANKS2; ankyrin repeat and sterile alpha motif domain containing 1B; ankyrin repeat and sterile alpha motif domain-containing protein 1B; C030032C09Rik; cajalin 2; cajalin-2; E2A-PBX1-associated protein; E530015N03Rik; EB1; EB-1; ENSMUSG00000078431; Gm10937; Gm1555; MGC26087; RGD1565556 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56899 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7Z6G8 | |
| ANKS1B | |
| Synthetic peptide directed towards the C-terminal of human ANKS1B (aa 1084-1133). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction