missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKS3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24058-1-AP-150UL
This item is not returnable.
View return policy
Description
Two isoforms of ANKS3 exist due to alternative splicing events. This antibody may recognize both of them.
The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins, proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also called activator 1, is a protein complex consisting of five distinct subunits of 145, 40, 38, 37, and 36.5 kD. This gene encodes the 40 kD subunit, which has been shown to be responsible for binding ATP. Deletion of this gene has been associated with Williams syndrome. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
| ANKS3 | |
| Polyclonal | |
| Unconjugated | |
| ANKS3 | |
| ANKS3, KIAA1977 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 124401, 72615 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6ZW76, Q9CZK6 | |
| Anks3 | |
| ANKS3 Fusion Protein Ag21252 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction