Learn More
Invitrogen™ Ankyrin B Monoclonal Antibody (N105/13), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545526
Description
1 μg/mL of MA5-45526 was sufficient for detection of Ankyrin-B in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 200kDa. No cross reactivity against Ankyrin-G. This antibody was formerly sold as clone S105-13.
This gene encodes a member of the ankyrin family of proteins that link the integral membrane proteins to the underlying spectrin-actin cytoskeleton. Ankyrins play key roles in activities such as cell motility, activation, proliferation, contact and the maintenance of specialized membrane domains. Most ankyrins are typically composed of three structural domains: an amino-terminal domain containing multiple ankyrin repeats; a central region with a highly conserved spectrin binding domain; and a carboxy-terminal regulatory domain which is the least conserved and subject to variation. The protein encoded by this gene is required for targeting and stability of Na/Ca exchanger 1 in cardiomyocytes. Mutations in this gene cause long QT syndrome 4. Multiple transcript variants encoding different isoforms have been described.
Specifications
| Ankyrin B | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q01484, Q8C8R3 | |
| ANK2 | |
| Synthetic peptide amino acids 203-496 of human Ankyrin-B. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N105/13 | |
| APC | |
| ANK2 | |
| AI835472; Ank2; ANK-2; Ank3; AnkB; ankyrin 2, brain; ankyrin 2, neuronal; ankyrin 3, epithelial; ankyrin B; ankyrin, brain; ankyrin-2; ankyrin-2, nonerythrocytic; ankyrin-B; AW491075; Brain ankyrin; brank-2; LQT4; Non-erythroid ankyrin | |
| Mouse | |
| Protein G | |
| RUO | |
| 109676, 287, 362036 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.