missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A1 Antibody (ANXA1/3869R), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08709PCP
This item is not returnable.
View return policy
Description
Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Annexin A1 | |
| Monoclonal | |
| PerCP | |
| annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
| Recombinant full-length human Annexin A1 protein (Uniprot: P04083) | |
| 0.1 mL | |
| Protein Kinase, Signal Transduction | |
| 301 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| ANXA1/3869R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction