missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A1 Antibody (OTI3A8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70174H
This item is not returnable.
View return policy
Description
Annexin A1 Monoclonal specifically detects Annexin A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.
Specifications
| Annexin A1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ANXA1 | |
| Full length recombinant protein of human Annexin A1 (NP_000691) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoassay | |
| OTI3A8 | |
| Western Blot, Immunohistochemistry, Knockout Validated | |
| annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase, Signal Transduction | |
| 301.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction