missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A1 Antibody (OTI3A8), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70174JF646
This item is not returnable.
View return policy
Description
Annexin A1 Monoclonal specifically detects Annexin A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| Annexin A1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ANXA1 | |
| Full length recombinant protein of human Annexin A1 (NP_000691) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| OTI3A8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
| annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase, Signal Transduction | |
| 301.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction