Learn More
Annexin A1 Antibody (SAIC-13B-19) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20092-0.2mg
 View more versions of this product
Product Code. 18625574
  • € 635.00 / 0.20mg
Description
Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Mass Spectrometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Annexin A1
Monoclonal
1 mg/mL
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, Immunofluorescence, Mass Spectrometry
SAIC-13B-19
Unconjugated
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein
Peptide 'AAYLQETGKPLDETLK' derived from Annexin A1 conjugated to KLH.
0.2 mg
Protein Kinase, Signal Transduction
301
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
IgG κ