missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A10 Antibody (OTI4D8), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71683AF488
This item is not returnable.
View return policy
Description
Annexin A10 Monoclonal specifically detects Annexin A10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Annexin A10 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ANXA10 | |
| Full length recombinant protein of human Annexin A10 (NP_009124) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI4D8 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| annexin 14, annexin A10, annexin-10, annexin-14, ANX14Annexin-10 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 11199.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction