Learn More
Annexin A10 Mouse anti-Human, Unconjugated, Clone: 1A7A11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66869-1-IG-20UL
 View more versions of this product
Product Code. 16849135
  • € 145.00 / 20µL
Description
The Annexin A10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Annexin A10. This antibody recognizes human, pig, mouse antigen. The Annexin A10 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Annexin A10
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1A7A11
Unconjugated
20 μL
11199