Learn More
Annexin A10 Mouse anti-Human, Unconjugated, Clone: 1A7A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66869-1-IG-20UL
Description
The Annexin A10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Annexin A10. This antibody recognizes human, pig, mouse antigen. The Annexin A10 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Annexin A10 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1A7A11 | |
| Unconjugated | |
| 20 μL | |
| 11199 | |
| IgG2a |