missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Annexin A10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15770101 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15770101 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15770101 Supplier Invitrogen™ Supplier No. PA552151

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PPLYDAHELW HAMKGVGTDE NCLIEILASR TNGEIFQMRE AYCLQYSNNL QEDIYSETSG HFRDTLMNLV QGTREEGYTD PAMAAQDAMV LWEACQQKTG EHKTMLQMIL CNK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The function of this gene has not yet been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANXA10
Gene Accession No. Q9UJ72
Gene Alias annexin 14; annexin A10; Annexin-10; annexin-14; ANX14; Anxa10; RGD1560956
Gene Symbols ANXA10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Annexin A10. Recombinant protein control fragment (Product #RP-88820).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11199
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.