missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A2 Antibody (ANEX 3D5/4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-2724-0.025mg
This item is not returnable.
View return policy
Description
Annexin A2 Monoclonal specifically detects Annexin A2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunoassay.
Specifications
| Annexin A2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunoassay | |
| ANXA2 | |
| Mixture of native membrane proteins from human bone tissues which would include whole native annexin proteins | |
| Protein A purified | |
| RUO | |
| Primary | |
| ANEX 3D5/4 detects native protein. Recommended concentration range: 1-10 ug/ml. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot | |
| ANEX 3D5/4 | |
| Unconjugated | |
| annexin A2, Annexin II, annexin-2, ANX2, ANX2L4LPC2, CAL1H, Calpactin I heavy chain, calpactin I heavy polypeptide, Calpactin-1 heavy chain, chromobindin 8, Chromobindin-8, LIP2PAP-IV, Lipocortin II, LPC2DP36, p36, Placental anticoagulant protein IV, Protein I | |
| Mouse | |
| 38 kDa | |
| 0.025 mg | |
| Cancer | |
| 302 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction