missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A2 Antibody (ANEX 3D5/4), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2724JF549
This item is not returnable.
View return policy
Description
Annexin A2 Monoclonal specifically detects Annexin A2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| AnnexinA2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ANXA2 | |
| Mixture of native membrane proteins from human bone tissues which would include whole native annexin proteins | |
| 0.1 mL | |
| Primary | |
| ANEX 3D5/4 detects native protein. Recommended concentration range: 1-10 ug/ml. | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| ANEX 3D5/4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| annexin A2, Annexin II, annexin-2, ANX2, ANX2L4LPC2, CAL1H, Calpactin I heavy chain, calpactin I heavy polypeptide, Calpactin-1 heavy chain, chromobindin 8, Chromobindin-8, LIP2PAP-IV, Lipocortin II, LPC2DP36, p36, Placental anticoagulant protein IV, Protein I | |
| Mouse | |
| Protein A purified | |
| Cancer | |
| 302 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction