Learn More
Invitrogen™ Annexin A2 Monoclonal Antibody (M298)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547646
Description
This antibody detects a 36 kDa* protein corresponding to the molecular mass of Annexin A2 on SDS-PAGE immunoblots of rat PC12 cells. The antibody has been shown to label Annexin A2 in immunocytochemical analysis of PC12 cells and immunohistochemical analysis of mouse diaphragm.
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| Annexin A2 | |
| Monoclonal | |
| 0.125 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P07355, P07356, Q07936 | |
| ANXA2 | |
| Clone M009 was generated from a proprietary antigen related to human Annexin A2 in MDA-MB-231 breast cancer cell line. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| M298 | |
| Unconjugated | |
| ANXA2 | |
| 33-kDa calcimedin; 33-kDa lymphocyte Ca>2+>- binding protein; 33kDa lymphocyte Ca>2+>- binding protein; 36-kDa calelectrin; annexin 2; annexin A2; annexin A2 isoform 2; annexin II; Annexin-2; ANX2; ANX2L4; ANXA2; AW215814; CAL1H; calpactin I heavy chain; calpactin I heavy chain (p36); calpactin I heavy polypeptide; calpactin-1 heavy chain; chromobindin 8; chromobindin-8; epididymis secretory protein Li 270; HEL-S-270; LIP2; lipocortin II; LPC2; LPC2D; p36; PAP-IV; Placental anticoagulant protein IV; protein I | |
| Mouse | |
| Protein A | |
| RUO | |
| 12306, 302, 56611 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |