missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Annexin A3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15934365
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15934365 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15934365 Supplier Invitrogen™ Supplier No. PA578780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human CACO-2 whole cell, human Hela whole cell, human A431 whole cell, human SH-SY5Y whole cell, rat lung tissue, mouse lung tissue. IHC: Mouse Cardiac Muscle tissue, Rat Cardiac Muscle tissue, Human Mammary Cancer tissue IHC-F: Mouse Cardiac Muscle Tissue, Rat Cardiac Muscle Tissue, Human Placenta Tissue. ICC/IF: A549 cell.

This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A3
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene ANXA3
Gene Accession No. O35639, P12429, P14669
Gene Alias 35-alpha calcimedin; Annexin 3; annexin A3; Annexin III; Annexin III (Lipocortin III); annexin III (lipocortin III, 1,2-cyclic-inositol-phosphate phosphodiesterase, placental anticoagulant protein III, calcimedin 35-alpha); Annexin3; annexin-3; ANX3; ANXA 3; ANXA3; calcimedin 35-alpha; inositol 1,2-cyclic phosphate 2-phosphohydrolase; LC3; Lipocortin III; LRRGT00047; PAP-III; Placental anticoagulant protein III
Gene Symbols ANXA3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human Annexin A3 (153-184aa RKALLTLADGRRDESLKVDEHLAKQDAQILYK).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11745, 25291, 306
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.